PD20047S0
Petri dish with Petri-Pad
sterile, Single use, diam. 47 mm, pkg of (150 units), For microbial analysis using 2 ml liquid medium
Synonym(s):
Petri dish with Pad for microbial membrane filtration
About This Item
Recommended Products
material
cellulose pad(s)
clear device
polystyrene
Quality Level
sterility
sterile
feature
ready-to-use
manufacturer/tradename
Petri-Pad™
packaging
pkg of (150 units)
technique(s)
sterile filtration: suitable
diam.
47 mm
color
clear
input
sample type cosmetics
sample type beer
sample type food(s)
sample type sport drink
sample type soft drink
sample(s) (environmental)
sample type cider
sample type beverage(s)
sample type bottled water
sample type wine
application(s)
bioburden testing
(Beverages, Water)
compatibility
Membrane filtration
shipped in
ambient
Related Categories
General description
Application
Features and Benefits
- Squared-off edges for one-handed opening
- Easily stackable
- Snug-fitting lids to prevent dehydration of media
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service