Skip to Content
Merck
All Photos(1)

Documents

P83068

Millipore

Power Cord for UK, Ireland, Hong Kong, Singapore

for use with Integritest®, Integritest®

Synonym(s):

IntegriTest Power Cord, Power cable

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

AC/DC input

100 - 200 V AC, 50 - 60 Hz

Quality Level

manufacturer/tradename

Integritest®

compatibility

for use with Integritest®

storage temp.

room temp

General description

Power Cord is used to power the IntegriTest System in UK, Ireland, Hong Kong and Singapore with 100 - 240 V, and 50-60 Hz

Application

Power Cord is used to power the IntegriTest System in UK, Ireland, Hong Kong and Singapore with 100 - 240 V, and 50-60 Hz

Legal Information

INTEGRITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service