Skip to Content
Merck
All Photos(1)

Key Documents

OP14

Sigma-Aldrich

Anti-c-ErbB2/c-Neu (Ab-2) Mouse mAb (9G6)

liquid, clone 9G6, Calbiochem®

Synonym(s):

Anti-ErbB2, Anti-Erythroblastosis Virus, Anti-HER2, Anti-c-Neu

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€354.00

€354.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€354.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€354.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9G6, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

Anti-c-ErbB2/c-Neu (Ab-2), mouse monoclonal, clone 9G6, recognizes the ~185 kDa c-ErbB2/c-Neu SK-BR-3 cells and breast carcinoma. It is validated for IP and IHC on frozen and paraffin sections.
Recognizes the ~185 kDa c-ErbB2/c-Neu protein in SK-BR-3 cells and breast carcinoma tissue. Does not cross-react with v-ErbB2 or the EGF receptor.
This clone expresses the kappa light chain isotype. Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0 Ag14 myeloma cells. Recognizes the ~185 kDa c-ErbB2/c-neu protein.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Negative Control
Normal skin tissue or HepG2 cells

Other Notes

Immunoprecipitates the 185 kDa c-neu protein and has been reported to stain formalin-fixed paraffin-embedded tissue sections of human breast carcinomas overexpressing the c-neu protein (see application references). This antibody reacts with a cell surface epitope of c-neu but does not cross-react with the EGF receptor. In one study ductal carcinomas in situ displaying large-cell, comedo growth type, were stained while none of 16 ductal carcinomas in situ of small-cell, papillary, or cribiform growth type were stained by this antibody. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prajakta S Oak et al.
International journal of cancer, 131(12), 2808-2819 (2012-04-19)
A major obstacle in the successful treatment of cancer is the occurrence of chemoresistance. Cancer cells surviving chemotherapy and giving rise to a recurrence of the tumor are termed cancer stem cells and can be identified by elevated levels of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service