Skip to Content
Merck
All Photos(1)

Key Documents

NZ-2ML-RU

Millipore

Reagent For Research Use - Rare Specificities & AHG

BIOSCOT Anti-k from the clone P3A118OL67, Human IgG Antibody, Blood Grouping Reagent for Research Use

Synonym(s):

Anti-k (clone P3A118OL67), Blood Grouping Reagent for Research Use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102

biological source

human

antibody form

unpurified

clone

P3A118OL67, monoclonal

form

liquid

packaging

pkg of 2 mL

concentration

1-5 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6-8

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide. Filtered through a 0.22μm terminal filter. Shelf Life: 36 months from date of filtration, For Non Clinical In Vitro research use only.

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service