MF0HC027H1
Millistak+® Pod Depth Filter
F0HC, filtration area 0.027 m², Lab Scale
Synonym(s):
Millistak+ Pod Depth Filter, F0HC media series, 0.027 m2 surface area, 1/pack
About This Item
Product Name
Millistak+® Pod Depth Filter, F0HC, cellulose, filtration area 0.027 m2, hydrophilic
material
cellulose
polypropylene
Quality Level
description
Secondary clarification of pretreated harvest by acid precipitation or flocculation, E. coli and yeast
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
2.1 bar max. differential pressure (30 psid) at 37 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 37 °C (Reverse)
2.1 bar max. differential pressure (30 psid) at 4 °C (Forward)
H
14 cm (5.3 in.)
L
22 cm (8.5 in.)
filtration area
0.027 m2
impurities
<0.25 EU/mL (LAL test, Aqueous extraction)
total organic carbon (TOC) residue
460-3200 ppb
input
sample type liquid
conductivity
21.4 – 42.7 μS/cm
filter grade
F0HC
shipped in
ambient
Related Categories
General description
This F0HC media series delivers optimal performance through a gradient density matri, ideal for secondary clarification post–centrifugation or pre-treated centrate applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Application
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Millistak+ HC dual-action media improves prefiltration and compresses clarification
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Preparation Note
Integrity is maintained after 2 autoclave cycles of 60 minutes @ 123 °C
Analysis Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service