MD0HC01FS1
Millistak+® HC Pod Depth Filter
D0HC, filtration area 0.11 m², Process Scale
Synonym(s):
Millistak+ HC Pod Depth Filter, D0HC media series, 0.11 m 2 surface area
About This Item
Recommended Products
Product Name
Millistak+® HC Pod Depth Filter, D0HC, cellulose, filtration area 0.11 m2, hydrophilic
material
cellulose
filter aid (inorganic)
glass fiber-reinforced polypropylene
Quality Level
description
Primary clarification directly out of the bioreactor
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
1 bar max. differential pressure (15 psid) at 80 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psi) at 25 °C
H
32 cm (12.5 in.)
L
62 cm (24.2 in.)
filtration area
0.11 m2
impurities
<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
gravimetric extractables
39-69 mg/m2
total organic carbon (TOC) residue
720-1260 ppb
input
sample type liquid
conductivity
8.66-10.5 μS/cm at 25 °C (product water)
filter grade
D0HC
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This D0HC media series delivers optimal performance through a gradient density matri, ideal for primary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Application
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Millistak+ HC dual-action media improves prefiltration and compresses clarification
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Sodium: <3.0 mg/ft2
Calcium: <0.7 mg/ft2
Magnesium: <0.2 mg/ft2
Mercury: <0.01 mg/ft2
Chromium: <0.01 mg/ft2
Aluminum: <0.01 mg/ft2
Potassium: <0.01 mg/ft2
Lead: <0.01 mg/ft2
Arsenic: <0.01 mg/ft2
Cobalt: <0.01 mg/ft2
Iron: <0.01 mg/ft2
Manganese: <0.01 mg/ft2
Titanium: <0.01 mg/ft2
Zinc: <0.01 mg/ft2
Preparation Note
1 autoclave cycle of 60 min @ 123 °C
Analysis Note
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service