Skip to Content
Merck
All Photos(2)

Documents

MCCHEAD01

Millipore

Microfil® V/S Head

Milliflex® Plus Pump, suitable for bioburden testing, water monitoring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel head

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

input

sample type food(s)
sample type beverage(s)

application(s)

bioburden testing
water monitoring

clinical
environmental
food and beverages
industrial qc

compatibility

Milliflex® Plus Pump

shipped in

ambient

General description

The Microfil® V/S head is designed to improve ease-of-use, reduce maintenance, and limit the risk of contamination. A continuous rim allows easy and safe membrane transfer with forceps, reducing the risk of touching the membrane filtration area. The porous filtration support can be removed simply by pressing down anywhere around its edge. The Microfil® V/S head is compatible with the Milliflex® Plus pump.

Application

Microfil® V/S head is an accessory of the Milliflex® Plus Pump used during filtration of the test samples for bioburden testing, and water monitoring.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service