MABX7132-10KC
Anti-hu N-term proBNP (NT-proBNP) clone 1312
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
antibody form
purified antibody
Quality Level
clone
13G12
monoclonal
purified by
using Protein A
species reactivity
human, mouse
concentration
(Please refer to the Certificate of Analysis for the lot-specific concentration.)
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG2a
General description
proBNP
Application
Confirmed matched pairing. Capture Antibody MABX7132-1KC; Detector Antibody MABX7154-1KC
Physical form
PBS (pH 7.4) with 0.09% sodium azide
Storage and Stability
Stable for 12 months at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service