Skip to Content
Merck
All Photos(1)

Documents

MABX4101

Sigma-Aldrich

Anti-human fatty acid binding protein (FABP) clone 101

clone 101, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

101, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

Gene Information

human ... FABP1(2168)

General description

Antibody recognizes Fatty Acid Binding Protein (FABP). FABP is a cytoplasmic protein involved in fatty acid metabolism. Measurement of heart type FABP levels in the blood can be used to assist in the diagnosis of certain cardiac conditions

Specificity

FABP

Application

Confirmed matched pairing. Capture Antibody MABX4028-1KC; Detector Antibody MABX4101-1KC
Detect FABP using this mouse monoclonal Anti-human fatty acid binding protein (FABP) clone 101 Antibody, Cat. No. MABX4101-1KC, validated for use in and Western Blotting and ELISA.
Research Category
Signaling

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service