Skip to Content
Merck
All Photos(3)

Key Documents

MABT1514

Sigma-Aldrich

Anti-Tenascin-R Antibody, clone 23-14

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€144.00
100 μG
€365.00

€144.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€144.00
100 μG
€365.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€144.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

23-14, monoclonal

mol wt

calculated mol wt 148.28 kDa
observed mol wt ~170 kDa

purified by

using protein G

species reactivity

human, mouse, chicken, rat

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

chicken ... TNR(396213)

General description

Tenascin-R (UniProt: Q00546; also known as TN-R, Restrictin) is encoded by the TNR gene (Gene ID: 396213) in chicken. Tenascins are a family of four large glycoproteins (Tenascin-C, R, X, and W) that share a cysteine-rich N-terminus, a sequence of fibronectin type III domains, and homologies to fibronectin-b at the C-terminus. Tenascin-R (TN-R), the smallest member of the tenascin family that can exist as homodimer or homotrimer. It interacts with different cells and matrix components and is involved in neural cell adhesion and neurite formation. Its interaction with contactin-1 enhances neurite outgrowth. It is expressed weakly in E6 embryo and reaches a maximum at E16 and declines in the adult. Its expression is observed in oligodendrocytes during myelination, and it accumulates at Ranvier nodes and by a subpopulation of neurons, including cerebellar stellate/basket cells, motoneurons in the spinal cord, and hippocampal interneurons. TN-R is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to generate the mature form. Its N-terminal region contains a cysteine-rich segment that it shares with other members of tenascin family. Within this segment there is a stretch of 20 amino acids that form three heptad repeats of hydrophobic amino acids in an a-helix that might generate a triple-stranded coiled coil to link three or two TN-R polypeptides to trimers or dimers. It also contains four EGF-like domains and nine fibronectin type-III (Fn-III) domains. Fn-III 9 mediates cell attachment and Fn-III 2-5 are reported to mediate neurofascin binding. It also contains a fibrinogen C-terminal domain (aa 1124-1339) that mediates its interaction with chondroitin Sulfate Proteoglycan 5. (Ref.: Nörenberg, U., et al. (1995). J. Cell Biol. 130(2); 473-484; Rathjen, FG., et al. (1991). Development.113(1); 151-164).

Specificity

Clone 23-14 is a mouse monoclonal antibody that detects Tenascin-R. It targets an epitope within the Fibronectin type-III 8 domain from the C-terminal half.

Immunogen

Immunoaffinity isolate of the monoclonal antibody F11 from detergent extracts of adult chicken brain plasma membrane preparations.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysates.

Western Blotting Analysis: A 1:125 dilution of this antibody detected Tenascin-R in Rat brain tissue lysates.

Tested Applications

Inhibition Assay: A representative lot of this antibody inhibited neural cell and retinal cell attachment to Tenascin-R. (Rathjen, F.G., et al. (1991). Development. 113(1):151-64; Norenberg, U., et al. (1995). J Cell Biol. 130(2):473-84; Norenberg, U., et al. (1996). Int J Dev Neurosci. 14(3):217-31).

Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution from a representative lot detected Tenascin-R in mouse brain and rat brain tissue sections.

Western Blotting Analysis: A representative lot detected Tenascin-R in Western Blotting applications (Wintergerst, E.S., et al. (2001). J Neurocytol. 30(4):293-301).

Immunohistochemistry Analysis: A representative lot detected Tenascin-R in Immunohistochemistry applications (Haunso, A., et al. (2000). Brain Res.;864(1):142-5; Wintergerst, E.S., et al. (2001). J Neurocytol.;30(4):293-301; Geissler, M., et al. (2013). J Neurosci.;33(18):7742-55).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Tenascin-R, clone 23-14, Cat. No. MABT1514, is a mouse monoclonal antibody that detects Tenascin-R and is tested for use in Immunohistochemistry (Paraffin), Inhibition Assay, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service