Skip to Content
Merck
All Photos(2)

Key Documents

MABT124

Sigma-Aldrich

Anti-Leukemia-associated RhoGEF/LARG Antibody, clone 13D2.1

clone 13D2.1, from mouse

Synonym(s):

Rho guanine nucleotide exchange factor 12, Leukemia-associated RhoGEF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13D2.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The protein named Leukemia-associated RhoGEF (LARG) or alternatively known as Leukemia-associated RhoGEF protein, and encoded by the gene ARHGEF12/KIAA0382/LARG, plays a role in the regulation of RhoA GTPase by guanine nucleotide-binding alpha-12 (GNA12) and alpha-13 (GNA13) proteins. Leukemia-associated RhoGEF (LARG) acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase and may act as GTPase-activating protein (GAP) for GNA12 and GNA13. Additionally, Leukemia-associated RhoGEF (LARG) interacts with RHOA, PLXNB1 and PLXNB2, and it interacts with IGF1R beta subunit and GCSAM proteins. Leukemia-associated RhoGEF (LARG) is localized to the cytoplasm and membrane. The protein translocates to the membrane upon stimulation. Leukemia-associated RhoGEF (LARG) widely expressed with isoform specific isoforms being found in certain tissues only, such as isoform 2 is primarily found in the intestine and testis.

Immunogen

Epitope: RGSL Domain
GST-tagged recombinant protein corresponding to the RGSL domain of human Leukemia-associated RhoGEF/LARG.

Application

Anti-Leukemia-associated RhoGEF/LARG Antibody, clone 13D2.1 is a highly specific mouse monoclonal antibody, that targets Leukemia-associated RhoGEF & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Leukemia-associated RhoGEF/LARG in HepG2 and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Leukemia-associated RhoGEF/LARG in 200 µg of HUVEC cell lysate.

Target description

~ 173 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service