Skip to Content
Merck
All Photos(1)

Key Documents

MABS469

Sigma-Aldrich

Anti-IA-2, clone 76F Antibody

clone 76F, from mouse

Synonym(s):

Receptor-type tyrosine-protein phosphatase-like N, R-PTP-N, Islet cell antigen 512, ICA 512, Islet cell autoantigen 3, PTP IA-2, PTPRN

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€485.00

€485.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€485.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€485.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

76F, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRN(5798)

General description

ICA 512, also known as Receptor-type tyrosine-protein phosphatase-like N (R-PTP-N), or Islet cell antigen 512 (ICA 512) or Islet cell autoantigen 3, or PTPIA-2, or IA-2, and encoded by the gene PTPRN/ICA3/ICA512, is a required protein for endocrine and neuroendocrine secretory processes. ICA 512 may be involved in processes specific for neurosecretory granules, such as their biogenesis, trafficking and regulated exocytosis, or ICA 512 may have a more general role in neuroendocrine functions. ICA 512 interacts with phosphorylated syntrophin beta 2/Distrophin, protecting it from protein cleavage by calpain I. Dephosphorylation of Distrophin upon insulin stimulation disrupts the interaction and results in its cleavage. ICA 512 is a membrane protein and found within neuroendocrine secretory granules and endocrine tissues. ICA 512 is expressed in brain, pituitary and gut tissues including the pancreas. ICA 512 is also now used as a diabetes-specific islet autoantigen biomarker in type diabetes patients.

Immunogen

Epitope: This antibody recognizes residues 626-630 found in juxtamembrane region of human and mouse IA-2, but not in IA-2b.
His-tagged recombinant protein corresponding to the residues 626-630 found in juxtamembrane region of human and mouse IA-2, but not IA-2b of human IA-2.

Application

Anti-IA-2 antibody, clone 76F is an antibody against IA-2 for use in western blotting, IHC & IP.
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected IA-2 in rat pancreas tissues and BTC3 cells (Piquer, S., et al. (2006). J Autoimmun. 26(3):215-222.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated IA-2 from recombinant rat, mouse, and human IA-2 (Piquer, S., et al. (2006). J Autoimmun. 26(3):215-222.).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in human pancreas tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected IA-2 in 10 µg of human pancreas tissue lysate.

Target description

~65 kDa observed. The full length protein is cleaved to yield a 65 kDa mature protein form that contains a lumenal domain, and an intracellular cytoplasmic domain characterized by a juxtamembrane region (Piquer, S., et al. (2006). J Autoimmun. 26(3):215-222.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aygul Subkhangulova et al.
eLife, 12 (2023-09-11)
Tomosyn is a large, non-canonical SNARE protein proposed to act as an inhibitor of SNARE complex formation in the exocytosis of secretory vesicles. In the brain, tomosyn inhibits the fusion of synaptic vesicles (SVs), whereas its role in the fusion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service