Skip to Content
Merck
All Photos(1)

Documents

MABS2217

Sigma-Aldrich

Anti-Syntaxin 5 Antibody

purified antibody, clone 1G11, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G11, monoclonal

mol wt

calculated mol wt 34 kDa
observed mol wt ~40 kDa

species reactivity

rat

species reactivity (predicted by homology)

mouse, human, bovine

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

Clone 1G11 is a mouse monoclonal antibody that detects Syntaxin-5. It targets an epitope within the cytoplasmic domain in the N-terminal half. It detects both isoform 1 and 2.

Immunogen

GST-tagged recombinant fragment corresponding to 83 amino acids from the C-terminal half, SNARE motif.

Application

Quality Control TestingEvaluated by Western Blotting in Rat brain tissue lysate.Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Syntaxin-5 in Rat brain tissue lysate.Tested ApplicationsWestern Blotting Analysis: A representative lot detected Syntaxin-5 in Western Blotting applications (Williams, A.L., et. al. (2004). Mol Biol Cell. 15(1):162-75).Immunocytochemistry Analysis: A representative lot detected Syntaxin-5 in Immunocytochemistry Blotting applications (Williams, A.L., et. al. (2004). Mol Biol Cell. 15(1):162-75).Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Syntaxin 5, clone 1G11, Cat. No. MABS2217, is a mouse monoclonal antibody that detects Syntaxin 5 and is tested for use in Immunocytochemistry and Western Blotting.

Target description

Syntaxin-5 (UniProt: Q08851; also known as STX-5) is encoded by the Stx5 (also known as Stx5a) gene (Gene ID: 65134) in rat. Syntaxin-5 is a single-pass type IV membrane protein that localizes throughout the Golgi apparatus and is most abundant in the cis-most cisternae. It is found in the endoplasmic reticulum (ER)-Golgi intermediate compartment membrane. It has a cytoplasmic domain (aa 1-333), a transmembrane domain (aa 334-354), and a single amino acid vesicular domain (aa 355). Its expression has been reported in the brain, heart, spleen, lung, liver, kidney, and testis. Syntaxin-5 mediates ER to Golgi transport and together with vesicular transport factor p115 (USO1) and Golgi matrix protein GM130 (GOLGA2), it is involved in tethering and fusion at the cis-Golgi membrane to maintain the stacked and inter-connected structure of the Golgi apparatus. Syntaxin-5 is shown to protect cells against proteotoxicity and is involved in clearance of insoluble protein deposits. (Ref.: Babazadeh, R., et al. (2019). Cell Rep. 28(8); 2096-2110; Diao, A., et al (2008); J. Biol. Chem. 283(11); 6957-6967).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service