Skip to Content
Merck
All Photos(1)

Documents

MABS1922

Sigma-Aldrich

Anti-Src-Like-Adaptor 2 (SLA2) Antibody, clone Z30P1F12*F4

clone Z30P1F12*F4, from mouse

Synonym(s):

Modulator of antigen receptor signaling, MARS, Src-like adapter protein 2, SLAP-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Z30P1F12*F4, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SLA2(84174)

General description

Src-like-adapter 2 (UniProt: Q9H6Q3; also known as Modulator of antigen receptor signaling; MARS; Src-like adapter protein 2; SLAP-2) is encoded by the SLA2 (also known as SLAP2, C20orf156) gene (Gene ID 84174) in human. SLAP-2 is localized to the plasma membrane and intracellular vesicles, including late endosomal vesicles. The SLAP-2 protein contains an NH2-terminal myristoylation consensus sequence and SH3 and SH2 Src homology domains, but lacks a tyrosine kinase domain. It is predominantly expressed in immune system, including T cells and monocytes and at low levels in B cells, placenta, prostate, colon, and skin. SLAP-2 serves as an adapter protein that negatively regulates T-cell receptor (TCR) signaling and is shown to block T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. In antigen receptor-stimulated cells, SLAP-2 associates with several tyrosine phosphorylated proteins, including the ubiquitin ligase Cbl. Deletion of the carboxyl terminus of SLAP-2 is shown to block its function and abolish its association with Cbl. Overexpression of SLAP-2 in T and B cell lines impairs antigen receptor-mediated signaling events, including the up-regulation of CD69, NFAT promoter activation, and calcium influx. Ref.: Holland, SJ et al. (2001). J. Exp. Med. 194(9):1263-1276. Pandey, A., et al. (2002). J. Biol. Chem. 277(21):19131-19138.

Specificity

Clone Z30P1F12*F4 detects -Src-Like-Adaptor 2 (SLA2) protein in human by targeting a sequence in the C-terminal region.

Immunogen

A synthetic peptide corresponding to 8 aa from the C-terminal region of human Src-like adapter 2 protein.

Application

Detect Src-like-adapter 2 using this mouse monoclonal Anti-Src-Like-Adaptor 2 (SLA2) antibody, clone Z30P1F12*F4, Cat. No. MABS1922, validated for use in Immunohistochemistry (Paraffin).
Research Category
Signaling

Quality

Evaluated by Immunohistochemistry in human cervical cancer, pancreas, and tonsil tissue.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected Src-Like-Adaptor 2 (SLA2) in human cervical cancer, pancreas, and tonsil tissue.

Target description

28.6 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service