Skip to Content
Merck
All Photos(4)

Key Documents

MABN663

Sigma-Aldrich

Anti-STMN2 Antibody, clone L5/1

clone L5/1, from mouse

Synonym(s):

Stathmin-2, Superior cervical ganglion-10 protein, Protein SCG10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€445.00

€445.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€445.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€445.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

L5/1, monoclonal

species reactivity

rat, human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Stathmin-like 2 (STMN2) protein is a phospho-protein and functions in microtubule dynamics and signal transduction. Studies show STMN2 plays a regulatory role in neuronal growth and thought to be involved in osteogenesis. Reduced levels, i.e. reduction in expression, have been associated with Down′s syndrome and Alzheimer′s disease. Data also suggests that STMN2 may play a role in modulating membrane interaction with the cytoskeleton during neurite outgrowth.

Immunogen

Epitope: C-terminus
Linear peptide corresponding to the C-terminus of rat STMN2.

Application

Detect STMN using this mouse monoclonal antibody, Anti-STMN2 Antibody, clone L5/1 validated for use in western blotting, IHC & ICC.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected STMN2 in 10 µg of rat fetal brain (E16) tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected STMN2 in rat hippocampus tissue.
Immunocytochemistry Analysis: A representative lot detected STMN2 in cultured hippocampal neurons (Prof. J. Trimmer, University of Califnornia, Davis.).

Quality

Evaluated by Western Blotting in SH-SY-5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected STMN2 in 10 µg of SH-SY-5Y cell lysate.

Target description

~21 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Aoki et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(4), 1473-1480 (2009-02-21)
Of the various microtubule-associated molecules, beta-tubulin III has been reported to be closely associated with the therapeutic efficacy of taxane-based chemotherapy against ovarian cancer. Stathmin and microtubule-associated protein 4 (MAP4) have been reported to play an important role in microtubule
Meghna Kannan et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(44), E9308-E9317 (2017-10-29)
The family of WD40-repeat (WDR) proteins is one of the largest in eukaryotes, but little is known about their function in brain development. Among 26 WDR genes assessed, we found 7 displaying a major impact in neuronal morphology when inactivated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service