Skip to Content
Merck
All Photos(1)

Key Documents

MABN2592

Sigma-Aldrich

Anti-RBP3 Antibody, clone mAb5

Synonym(s):

IRBP, Interphotoreceptor retinoid-binding protein, Interstitial retinol-binding protein, Protein 7S, Retinol-binding protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

mAb5, monoclonal

species reactivity

bovine

packaging

antibody small pack of 100

technique(s)

electron microscopy: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

Gene Information

bovine ... RBP3(281443)

Specificity

Clone mAb5 is a mouse monoclonal antibody that detects Retinol-binding protein 3 (RBP3). It targets an epitope within the N-terminal half.

Immunogen

Purified Retinol-binding protein 3 (RBP3) from bovine retina.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from Bovine retinal tissue.

Western Blotting Analysis: A 1:500 dilution of this antibody RBP3 in lysate from Bovine retinal tissue.

Tested Applications

Electron Microscopy: A representative lot detected RBP3 in Electron Microscopy applications (Sears, A.E., et al. (2020). FASEB J. 34(10):13918-13934).

Immunoprecipitation Analysis: A representative lot immunoprecipitated RBP3 in Immunoprecipitation applications (Sears, A.E., et al. (2020). FASEB J. 34(10):13918-13934).

Western Blotting Analysis: A representative lot detected RBP3 in Western Blotting applications (Sears, A.E., et al. (2020). FASEB J. 34(10):13918-13934).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Retinol-binding protein 3 (UniProt: P12661; also known as Interphotoreceptor retinoid-binding protein, IRBP, Interstitial retinol-binding protein, RBP3, Protein 7S) is encoded by the RBP3 gene (Gene ID: 281443) in bovine. RBP3 is synthesized with a signal peptide (aa 1-17) and a propeptide (aa 18-22) that are subsequently cleaved off to produce the mature form. It is a lipophilic glycoprotein that is secreted by rod and cone photoreceptors and plays a major role in photoreceptor homeostasis as well as retinoid and polyunsaturated fatty acid transport between the neural retina and retinal pigment epithelium. Its expression is observed early in retinal differentiation. RBP3 shuttles 11-cis and all trans retinoids between the retinol isomerase in the pigment epithelium and the visual pigments in the photoreceptor cells of the retina. It can bind many retinoids and polyunsaturated fatty acids and in addition to retinoid binding, it also plays a role in retinal development and homeostasis. RBP3 also displays thiol-dependent antioxidant activity that maintains the delicate redox balance in the normal retina. RBP3 dysregulation has been linked to retinal diseases. Mice lacking RBP3 exhibit severe early and progressive photoreceptor degeneration that is characterized by a reduction in both length and numbers of cone sheaths. In mammals four protein subunits of RBP3 have been described (M1-4), each consisting of about 300 amino acids. Clone mAb5 has been mapped to M2 subunit of RBP3. (Ref.: Sears, AE., et al. (2020) FASEB J. 34(10); 13918-13934; Zheng, S., et al. (2020). Front. Cell. Neurosci. 14; Article 577935).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl and 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service