Skip to Content
Merck
All Photos(1)

Key Documents

MABN2484

Sigma-Aldrich

Anti-ErbB4 Antibody, clone mAB10

clone mab10, from rabbit

Synonym(s):

Receptor tyrosine-protein kinase erbB-4, Proto-oncogene-like protein c-ErbB-4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€354.00

€130.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€130.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mab10, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Erbb4(13869)

General description

Receptor tyrosine-protein kinase erbB-4 (UniProt: Q61527; also known as EC: 2.7.10.1, Proto-oncogene-like protein c-ErbB-4) is encoded by the Erbb4 (also known as Mer4) gene (Gene ID: 13869) in murine species. ErbB-4 is a single-pass type I membrane protein of the EGF receptor family that plays an essential role as cell surface receptor for neuregulins (NRG) and EGF family members. ErbB-4 is monomeric in the absence of bound ligand and it homo- or heterodimerizes with another ERBB family member upon ligand binding. ErbB-4 is autophosphorylated on tyrosine residues in response to ligand binding that triggers receptor dimerization and autophosphorylation at specific tyrosine residues, which then serve as binding sites for scaffold proteins and effectors. ErbB-4 is reported to regulate the development of the heart, the central nervous system, and the mammary gland. It is shown to be involved in gene transcription, cell proliferation, differentiation, migration, and apoptosis. It is expressed in the developing and adult cerebral cortex, and represents the major NRG receptor in central neurons and regulates a diverse array of neuronal processes, including migration, differentiation, neurotransmission, and synaptic plasticity. NRG-1 and ErbB-4 are essentials of a signaling pathway that is critical for the proper development of inhibitory cortical and hippocampal circuits. ErbB-4 expression is reported to be restricted to GABAergic neurons in the cerebral cortex where it plays a role in regulating GABAergic synaptic development and interneuron migration. ErbB-4 is synthesized with a signal peptide (aa 1-25), which is cleaved off in the mature form. The mature form has an extracellular domain (aa 26-652), a short transmembrane domain (aa 653-673), and a long cytoplasmic tail (aa 674-1308). Its protein kinase domain is localized in the cytoplasmic tail (aa 718-985). Three isoforms of ErbB-4 have been described that are produced by alternative splicing.

Specificity

Clone mAB10 is a rabbit monoclonal antibody that detects mouse Receptor tyrosine-protein kinase erbB-4. It targets an epitope with in the C-terminal region.

Immunogen

GST-tagged recombinant protein corresponding to 204 amino acids from the C-terminal region of murine Receptor tyrosine-protein kinase erbB-4.

Application

Anti-ErbB4, clone mAB10, Cat. No. MABN2484, is a highly specific rabbit monoclonal antibody that targets Receptor tyrosine-protein kinase ErbB-4 and has been tested for use in ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected ErbB4 in mouse embryo tissue sections.

Western Blotting Analysis: A representative lot detected ErbB4 in Western Blotting applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).

ELISA Analysis: A representative lot detected ErbB4 in ELISA applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).

Immunohistochemistry Analysis: A representative lot detected ErbB4 in Immunohistochemistry applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).

Immunoprecipitation Analysis: A representative lot immunoprecipitated ErbB4 in Immunoprecipitation applications (Vullhorst, D., et. al. (2009). J Neurosci. 29(39):12255-64).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse cerebral cortex tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected ErbB4 in mouse cerebral cortex tissue sections.

Target description

146.85 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rabbit monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service