Skip to Content
Merck
All Photos(2)

Key Documents

MABN2330

Sigma-Aldrich

Anti-Catenin delta-2 Antibody, clone 4H2.1

clone 4H2.1, from mouse

Synonym(s):

Delta-catenin, GT24, Neural plakophilin-related ARM-repeat protein, NPRAP, Neurojungin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€342.00

€130.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€342.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H2.1, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CTNND2(1501)

General description

Catenin delta-2 (UniProt: Q9UQB3; also known as Delta-catenin, GT24, Neural plakophilin-related ARM-repeat protein, NPRAP, Neurojungin) is encoded by the CTNND2 (also known as NPRAP) gene (Gene ID: 1501) in human. Delta Catenin is a component of the cadherin-catenin cell adhesion complex and is enriched in synaptosomes. Catenin Delta 2 is a member of the beta-catenin superfamily and is expressed in the brain where it is required for normal cognitive development and its highest expression is observed in fetal brain. Catenin Delta 2 is phosphorylated by Cdk5 and by GSK-3beta. Catenin Delta 2 plays a critical role in neuronal development, particularly in the formation and/or maintenance of dendritic spines and synapses. It is also involved in the regulation of Wnt signaling where it acts on beta-catenin turnover, facilitating beta-catenin interaction with GSK3beta, phosphorylation, ubiquitination, and degradation. When bound to ZBTB33 (Zinc Finger and BTB Domain Containing 33), it functions as a transcriptional activator. Catenin delta-2 also promotes the disruption of E-cadherin based adherens junction to favor cell spreading upon stimulation by hepatocyte growth factor. Two isoforms of Catenin delta-2 have been reported that are produced by alternative splicing. It has nine Armadillo (ARM) repeats and a coiled coil region (aa 49-84). Defects in CTNND2 gene are known to cause autism and delayed acquisition of psychomotor skills. (Ref.: Yuan, L et al. (2015). J. Biol. Chem. 290(17):10947-57).

Specificity

Clone 4H2.1 specifically detects Catenin delta-2 in human and murine cells. It targets an epitope with in the C-tereminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 145 amino acids from the C-terminal region of human Catenin delta-2.

Application

Anti-Catenin delta-2, clone 4H2.1 Antibody, Cat. No. MABN2330, is a highly specific mouse monoclonal antibody that targets Catenin delta-2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution from a representative lot detected Catenin delta-2 in rat thalamus, rat cerebellum, and human cerebral cortex tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 0.02 µg/mL of this antibody detected Catenin delta-2 in mouse cerebellum tissue lysate.

Target description

~150 kDa observed; 132.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service