MABN1848
Anti-DYRK1A Antibody, clone 8D9
clone 8D9, from mouse
Synonym(s):
Dual specificity tyrosine-phosphorylation-regulated kinase 1A, Dual specificity YAK1-related kinase, MNBH, Protein kinase minibrain homolog, RP86
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
8D9, monoclonal
species reactivity
rat, mouse, human
species reactivity (predicted by homology)
chicken (based on 100% sequence homology)
technique(s)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
chicken ... Dyrk1A(395233)
human ... DYRK1A(1859)
mouse ... Dyrk1A(13548)
rat ... Dyrk1A(25255)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A 1:500 dilution from a representative lot detected DYRK1A in 10 µg of HeLa cell lysate.
Western Blotting Analysis: A representative lot detected DYRK1A in NIH/3T3 cell lysate and human frontal cortex homogenates, as well as in DYRK1A and actin immunoprecipitates (Dowjat, K., et al. (2012). J. Neuropathol. Exp. Neurol. 71(12):1100-1112).
Western Blotting Analysis: A representative lot detected DYRK1A levels in lymphoblastoid cell lines (LCLs) from healthy control, Down syndrome (DS) and fragile X (FraX) human subjects (Dowjat, K., et al. (2012). J. Neuropathol. Exp. Neurol. 71(12):1100-1112).
Western Blotting Analysis: Representative lots detected DYRK1A in rat brain tissue homogenate and subfractionated extracts. The nuclear fraction had low DYRK1A, whereas the majority DYRK1A was found in the postnuclear fractions with clathrin-coated vesicles (CCVs) containing the highest level (Murakami, N., et al. (2012). PLoS One. 7(4):e34845; Murakami, N., et al. (2009). Biochemistry. 48(39):9297-9305).
Western Blotting Analysis: A representative lot detected recombinant rat DYRK1A-bound protein bands by a Western blotting-based DYRK1A overlay assay (Murakami, N., et al. (2009). Biochemistry. 48(39):9297-9305).
Western Blotting Analysis: A representative lot detected recombinant rat DYRK1A in GST-End1, but not GST-End2, pull-down. Clone 8D9 detected the C-terminal PEST domain deletion DYRK1A497 construct in the GST-amphiphysin SH3 domain (GST-Am-SH3) pull-down only in the presence of dynamin (Murakami, N., et al. (2009). Biochemistry. 48(39):9297-9305).
Neuroscience
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected DYRK1A in 10 µg of HEK293 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service