Skip to Content
Merck
All Photos(2)

Documents

MABN1587

Sigma-Aldrich

Anti-MATR3 Antibody, clone 3G1.1

clone 3G1.1, from mouse

Synonym(s):

Matrin-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G1.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MATR3(9782)

General description

Matrin-3 (UniProt P43243) is encoded by the MATR3 (also known as ALS21, KIAA0723, VCPDM) gene (Gene ID 9782) in human. Matrin-3 is an inner nuclear matrix protein involved in multiple nuclear processes, including DNA double strand break response, DNA replication/repair, chromatin organization, and RNA stability and processing. Matrin-3 contains two RNA recognition motifs (a.a. 398-473/RRM1 and a.a. 496-571/RRM2) that mediate RNA-binding and a DNA-binding zinc finger motif (a.a. 801-832). Matrin-3 is known to interact with the RNA-binding protein TDP-43 and MATR3 gene mutations are linked to amyotrophic lateral sclerosis (ALS). Matrin-3 is also a known target of multiple alpha herpes viruses and is known to be utilized by HIV-1 as a Rev cofactor.

Specificity

Expected to react with both spliced isoforms of human MATR3 reported by UniProt (P43243).

Immunogen

GST-tagged recombinant internal fragment from the N-terminal half of human martin-3.

Application

Immunohistochemistry Analysis: An 1:250 dilution from a representative lot detected MATR3 in human cerebellum, human small intestine, and mouse cerebral cortex tissue sections.
This Anti-MATR3 Antibody, clone 3G1.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of MATR3.

Quality

Evaluated by Western Blotting in SHSY-5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MATR3 in 10 µg of SHSY-5Y cell lysate.

Target description

~110 kDa observed. 94.62 kDa (isoform 1) and 62.58 kDa (isoform 2) calculated. The detected band size is consistent with the reported apparent molecular weight (Nakayasu, H., and Berezney, R. (1991). Proc. Natl. Acad. Sci. U. S. A. 88(22):10312-10316).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service