Skip to Content
Merck
All Photos(1)

Documents

MABN1544

Sigma-Aldrich

Anti-IP3R3 Antibody, clone 6G12.1

clone 6G12.1, from mouse

Synonym(s):

Inositol 1, 4, 5-trisphosphate receptor type 3, InsP3R3, IP3 receptor isoform 3, IP3R 3, Type 3 inositol 1, 4, 5-trisphosphate receptor, Type 3 InsP3 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6G12.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITPR3(3710)

General description

InsP3R3, IP3 receptor isoform 3, IP3R 3, Type 3 inositol 1,4,5-trisphosphate receptor, Type 3 InsP3 receptor) is encoded by the ITPR3 gene (Gene ID 3710) in human. The inositol 1,4,5-trisphosphate (InsP3) receptors, InsP3R1, InsP3R2, and InsP3R3, play an important role in maintaining normal epithelial secretory function by mediating intracellular Ca2+ release. InsP3R3 is the major isoform in hepatic bile ducts, where it is concentrated along the subapical portion of the ER and regulates bicarbonate (HCO3 ) and chloride (Cl ) secretion into the bile. Wherease hepatocytes express mainly InsP3R2 that is involved in modulating the secretion of bile solutes. A loss of InsP3R3 expression in cholangiocytes is found in primary biliary cirrhosis (PBC), an autoimmune liver disease characterized by the loss of bile ducts and impaired bile secretion. Likewise, InsP3R3 and InsP3R2 double knockout mice are reported to exhibit severe defects of pancreatic zymogen secretion. Human InsP3R3 is a 6-transmembrane (a.a. 2203-2223, 2236-2256, 2265-2285, 2326-2346, 2369-2389, 2497-2517) ER membrane protein, having both its N- and C-terminal ends (a.a. 1-2202 and 2518-2671) exposed at the cytoplasmic side. The N-terminal cytoplasmic region contains five MIR (Mannosyltransferase, InsP3R, RyR) domains (a.a. 113-173, 174-224, 232-288, 295-372, 378-434) and the InsP3-binding sites (a.a. 266-270, 507-510, 567-569).

Specificity

Clone 6G12.1 targets an epitope in the C-terminal half of human IP3R3 (IP3 Receptor 3) N-terminal cytoplasmic domain.

Immunogen

GST-tagged recombinant human IP3R3 (IP3 Receptor 3) N-terminal cytoplasmic domain fragment.

Application

Anti-IP3R3 Antibody, clone 6G12.1, Cat. No. MABN1544, is a highly specific mouse monoclonal antibody that targets ITPR3 and has been tested in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected IP3R3 (IP3 Receptor 3) in 10 µg of HeLa cell lysate.

Target description

~260 kDa observed. 304.1 kDa calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service