Skip to Content
Merck
All Photos(3)

Documents

MABN1524

Sigma-Aldrich

Anti-CHCHD10 Antibody, clone 11F11.2

clone 11F11.2, from mouse

Synonym(s):

Coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial, Protein N27C7-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11F11.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial (UniProt: Q8WYQ3; also known as Protein N27C7-4) is encoded by the CHCHD10 (also known as C22orf16) gene (Gene ID: 400916) in human. CHCHD10 belongs to a family of mitochondrial proteins located in the intermembrane space that is enriched at cristae junctions. CHCHD10 displays a ubiquitous distribution pattern, but its expression is highest in heart and liver and lowest in spleen. It is involved in the maintenance of mitochondrial organization, mitochondrial cristae structure, and mitochondrial DNA stability. CHCD10 has a non-structured N-terminal region, followed by a highly hydrophobic helix and a C-terminal CHCH domain characterized by a Cx(9)C motif and 2 additional cysteines. The four cysteine residues (102, 112, 122 and 132) of the CHCH domain are involved in two disulphide bonds. Overexpression of a CHCHD10 mutant allele in HeLa cells is reported to lead to fragmentation of the mitochondrial network and ultrastructural major abnormalities including loss, disorganization and dilatation of cristae. Mutations in CHCHD10 gene are reported to cause frontotemporal dementia and/or amyotrophic lateral sclerosis 2 (FTDALS2). (Ref.: Bannwarth, S., et al. (2014). Brain. 2014 137(Pt 8):2329-2345).

Specificity

Clone 11F11.2 detects CHCHD10 in human brain and liver.

Immunogen

GST-tagged recombinat fragment corresponding to 79 amino acids from human CHCHD10.

Application

Anti-CHCHD10 antibody, clone 11F11.2, Cat. No. MABN1524, is a highly specific mouse monoclonal antibody, that targets CHCHD10 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CHCHD10 in 10 µg of human liver tissue lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CHCHD10 in human small intestine, human tonsil, human liver, and human prostate cancer tissue.


Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CHCHD10 in 10 µg of human brain tissue lysate.

Target description

~15 kDa observed; 14.15 kDa calulated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service