Skip to Content
Merck
All Photos(1)

Documents

MABF983

Sigma-Aldrich

Anti-Complement C3b/iC3b/C3dg Antibody, clone 1H8

clone 1H8, from mouse

Synonym(s):

Complement C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, Complement C3 beta chain, C3-beta-c, C3bc, Complement C3 alpha chain, C3a anaphylatoxin, Acylation stimulating protein, ASP, C3adesArg, Complement C3b alpha′ chain, Complem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H8, monoclonal

species reactivity

human, primate

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... C3(718)

Related Categories

General description

Complement C3 (UniProt P01024; also known as C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1) is encoded by the C3 (also known as CPAMD1) gene (Gene ID 718) in human. C3 is initially translated with an N-terminal 22-amino acid signal peptide sequence, which is then removed to produce the 1641-amino acid mature C3. It plays a central role in the activation of complement system. Its activation is required for the activation of both classical and alternative pathway of complement (CPC and APC, respectively). C3 is cleaved into C3a and C3b during CPC activation by the C3-convertase C4b2a composed of the activated C4 and C2. In APC, C3 is cleaved by the C3-convertase C3bBb composed C3b and the activated form of factor B (Bb). C3b serves as an opsonizing agent, and can be further cleaved by Factor I into C3c and C3d. iC3b is a proteolytically inactive C3b fragment that still opsonizes target microbes or cells, but cannot further amplify/activate the complement cascade through APC. iC3b can be further cleaved to C3dg, and finally to C3d. Unregulated activation of APC can result in paroxysmal nocturnal hemoglobinuria (PNH) that is characterized by chronic intravascular hemolysis. Clinical C5-neutralizing mAb treatment prevents the formation of cytolytic membrane attack complex (MAC) of complement, but does not block APC activation. Consequently, PNH patients are left with immune-mediated hemolytic anemia and their erythrocytes become opsonized with complement C3. Monoclonal antibodies (mAbs) against C3b/iC3b are useful for monitoring and studying C3b/iC3b deposit on PNH blood cells and mAbs with neutralizing activities are useful tools for studying C3-mediated CPC and APC.

Specificity

This clone binds C3b, iC3b, and C3dg, but does not block the activation of alternative pathway of complement (APC).

Immunogen

Epitope: iC3dg
Sepharose 4B beads with surface C3b/C3bi deposits via APC in normal human serum corresponding to the iC3dg of human Complement C3b/iC3b/C3dg.

Application

Flow Cytometry Analysis: A presentative lot detected C3b/iC3b/C3dg deposit on paroxysmal nocturnal hemoglobinuria (PNH) patient-derived erythrocyte ghosts due to alternative pathway of complement (APC) activation-mediated hemolysis in acidified normal human serum (aNHS; pH 6.4) (Lindorfer, M.A., et al. (2010). Blood. 115(11):2283-2291).
This mouse monoclonal Anti-Complement C3b/iC3b/C3dg Antibody, clone 1H8, Cat. No. MABF983 is validated for use in Flow Cytometry, for the detection of C3b.

Quality

Flow Cytometry Analysis: This antibody (200 ug mAb/5 x 10e6 cells/mL) detected C3b/iC3b/C3dg deposit on human Burkett′s lymphoma Raji B cells opsonized with anti-CD20 mAb Rituximab (RTX) in the presence of 50% normal human serum (NHS).

Target description

187 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service