MABF3074
Anti-SRF Antibody, clone 2C5
Synonym(s):
Serum response factor
About This Item
biological source
rat
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2C5, monoclonal
mol wt
calculated mol wt 51.25 kDa
purified by
using protein G
species reactivity
human, mouse
packaging
antibody small pack of 100 μg
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgG2aκ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
mouse ... Srf(20807)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Immunohistochemistry (Paraffin) in human psoriatic skin tissue sections.
Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:50 dilution of this antibody detected Serum response factor (SRF) in human psoriatic skin tissue sections.
Tested Applications
Immunofluorescence Analysis: A representative lot detected SRF in Immunofluorescence applications (Koegel, H., et al. (2009). J Clin Invest. 119(4):899-910).
Immunohistochemistry Applications: A representative lot detected SRF in Immunohistochemistry applications (Koegel, H., et al. (2009). J Clin Invest. 119(4):899-910).
Western Blotting Analysis: A representative lot detected SRF in Western Blotting applications (Koegel, H., et al. (2009). J Clin Invest. 119(4):899-910).
Chromatin Immunoprecipitation Analysis (ChIP): A representative lot detected SRF in Chromatin Immunoprecipitation applications (Hu, X., et al. (2019). Nat Commun. 10(1):1695).
Immunoprecipitation Analysis: A representative lot detected SRF in Immunoprecipitation applications (Hu, X., et al. (2019). Nat Commun. 10(1):1695).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service