Skip to Content
Merck
All Photos(1)

Key Documents

MABF3048

Sigma-Aldrich

Anti-MERS-CoV Nucleoprotein Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€129.00
100 μL
€324.00

€129.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€129.00
100 μL
€324.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€129.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

46, monoclonal

mol wt

calculated mol wt 45.05 kDa
observed mol wt ~45 kDa

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Nucleoprotein (UniProt: R9UM87; also known as Nucleocapsid protein, NC, Protein N) is encoded by the N gene (Gene ID: 14254601) in Middle East respiratory syndrome-related coronavirus (MERS-CoV). The MERS-CoV is a positive-strand RNA virus of the betacoronavirus genus within the Coronaviridae family that causes severe respiratory syndrome in human. The mature MERS-CoV contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). Nucleoprotein of MERS-CoV is a homooligomeric multifunctional protein that plays roles in genome encapsidation, RNA synthesis during replication, host translational shutoff, and innate immunity disruption. It packages the positive strand viral genome RNA into helical ribonucleocapsid and is involved in virion assembly through its interactions with the viral genome and membrane protein M. The nucleoprotein is reportedly organized into two folded domains known as the N-terminal domain (NTD) and the C-terminal domain (CTD) that are separated by a disordered region (LKR) containing a serine/ arginine stretch. Clone 46 displays specificity for MERS-CoV nucleoprotein and does not show cross-reactivity with nucleoprotein from other related viruses, including SARS coronavirus. (Ref.: Nguyen, THV., et al. (2019). Acta Crystallogr. D. Struct. Biol. 75(1); 8-15; Yamaoka, Y., et al. (2016). Front. Microbiol. 7; Article 509).

Specificity

Clone 46 is a mouse monoclonal antibody that detects nucleoprotein from Middle East respiratory syndrome-related coronavirus (MERS-CoV).

Immunogen

His-tagged recombinant fragment corresponding to 292 amino acids from the nucleoprotein of Middle East respiratory syndrome-related coronavirus (MERS-CoV).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant MERS-CoV nucleoprotein.

Western Blotting Analysis: A 1:50,000 dilution of this antibody detected recombinant MERS-CoV Nucleoprotein.

Tested Applications


ELISA Analysis: A representative lot detected MERS-CoV Nucleoprotein in ELISA applications (Yamaoka, Y., et. al. (2016). Front Microbiol. 7:509).

Western Blotting Analysis: A representative lot detected MERS-CoV Nucleoprotein in Western Blotting applications (Yamaoka, Y., et. al. (2016). Front Microbiol. 7:509).

Immunoprecipitation Analysis: A representative lot immunoprecipitated MERS-CoV Nucleoprotein in Immunoprecipitation applications (Yamaoka, Y., et. al. (2016). Front Microbiol. 7:509).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MERS-CoV Nucleoprotein, clone 46, Cat. No. MABF3048, is a mouse monoclonal antibody that detects MERS-CoV Nucleoprotein and is tested for use in ELISA, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service