MABF3034
Anti-NKG2D/CD314 Antibody, clone A10
Synonym(s):
B-cell-activating factor, BAFF, CD257, Tumor necrosis factor ligand superfamily member 13B
About This Item
biological source
hamster
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
A10, monoclonal
species reactivity
mouse
packaging
antibody small pack of 100
technique(s)
flow cytometry: suitable
inhibition assay: suitable
isotype
IgGκ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
Gene Information
mouse ... Tnfsf13b(27007)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in C57BL/6 Mouse splenocytes.
Flow Cytometry Analysis (FC): 1.0 µg of this antibody detected NKG2D/CD314 in one million C57BL/6 mouse splenocytes.
Tested Applications
Flow Cytometry Analysis: A representative lot detected NKG2D/CD314 in Flow Cytometry applications (Ho, E.L., et al. (2002). J Immunol. 169(7):3667-75).
Agonist Function: A representative lot stimulated the production of GM-CSF by KY-1 NK clones and C57BL/6 LAK cells and also stimulated NK cells to kill Daudi targets. (Ho, E.L., et al. (2002). J Immunol. 169(7):3667-75).
Agonist Functon: A representative lot increased Vav phosphorylation in freshly isolated splenic C57BL/6 NK cells. (Ghasemi, R., et al. (2016). Nat Commun. 7:12878).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service