Skip to Content
Merck
All Photos(1)

Key Documents

MABF2786

Sigma-Aldrich

Anti-IL-9 Antibody, clone MH9A3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€146.00
100 μL
€367.00

€146.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€146.00
100 μL
€367.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

€146.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MH9A3, monoclonal

mol wt

calculated mol wt 15.91 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
flow cytometry: suitable
multiplexing: suitable
neutralization: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... il9> IL9(3578)

General description

Interleukin-9 (UniProt: P15248; also known as IL-9, Cytokine P40, T-cell growth factor P40) is encoded by the IL9 gene (Gene ID: 3578) in human. IL-9 is a pleiotropic cytokine that plays a significant role in tolerance and inflammation. It is expressed by a variety of immune cells. It is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to generate the mature form. It supports the growth of mast cells, T cells, and several other cell types, including erythroid progenitors, fetal thymocytes, human megakaryoblastic leukemic cells, and myeloid precursor cells. It is also reported to supports IL-2 independent and IL-4 independent growth of helper T-cells. It also plays a role in promoting IgE production by B cells. IL-9 actions are mediated via its binding to IL-9R that results in the formation the IL-9R heterocomplex. Following its binding to the IL-9R , the SHC is phosphorylated, which then binds to Grb2 that subsequently activates SoS, a GTP exchange factor for Ras GTPases. This results in the activation of Ras GTPase that activates Raf kinase followed by the activation of MEK1/2 and ERK1/2. Although IL-9 is reported to activate the MAPK pathway in several lymphoid and hematopoietic cells, this activation is weaker compared to other cytokines. Elevated levels of IL-9 and IL-9R are observed in lungs of asthmatic subjects. (Ref.: Chakraborty, S., et al. (2019). Int. J. Mol. Sci. 20(9); 2113).

Specificity

Clone MH9A3 is a mouse monoclonal antibody that detects human Interleukin-9.

Immunogen

Ovalbumin-conjugated recombinant human Interleukin-9.

Application

Quality Control Testing

Evaluated by ELISA with recombinant human IL-9.

ELISA Analysis: Various dilution of this antibody detected recombinant human Interleukin-9 (IL-9).

Tested Applications

ELISA Analysis: A representative lot detected IL-9 in ELISA applications (Jenmalm, M.C., et al. (2001). Clin Exp Allergy.;31(10):1528-35; Faulkner, H., et al. (2002). J Infect Dis.;185(5):665-72; Bottcher, M.F., et al. (2003). Pediatr Allergy Immunol.;14(5):345-50; Holz, L.E., et al. (2005). J Inflamm (Lond).;2:3.; Stephens, G.L., et al. (2011). Eur J Immunol.;41(4):952-62; Muls, N., et al. (2012). J Neuroimmunol.;243(1-2):73-80; Peters, C., et al. (2018). Oncoimmunology.;8(1):e1522471).

Flow Cytometry Analysis: A representative lot detected IL-9 in Flow Cytometry applications (Stephens, G.L., et al. (2011). Eur J Immunol.;41(4):952-62; Peters, C., et al. (2018). Oncoimmunology.;8(1):e1522471).

Multiplexing: A representative lot detected IL-9 in Multiplexing applications (Peters, C., et al. (2018). Oncoimmunology.;8(1):e1522471).

Neutralizing: A representative lot neutralized the action of IL-9 in Neutralizing applications (Stephens, G.L., et al. (2011). Eur J Immunol.;41(4):952-62).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-9, clone MH9A3, Cat. No. MABF2786, is a mouse monoclonal antibody that detects Interleukin-9 and is tested for use in ELISA, Flow Cytometry, Multiplexing, and Neutralizing.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service