Recommended Products
antibody form
purified antibody
clone
EBVCS4
monoclonal
purified by
(Ligatrap)
species reactivity
human, human, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
isotype
IgMκ
UniProt accession no.
General description
Specificity
Application
Immunofluorescence Analysis: A representative lot detected BLAST-2 (CD23) in Immunofluorescence applications (Kintner, C., et. al. (1981). Nature. 294(5840):458-60).
Immunoprecipitation Analysis: A representative lot immunoprecipitated BLAST-2 (CD23) in Immunoprecipitation applications (Sugden, B., et. al. (1983). J Virol. 46(3):800-7).
Quality
Flow Cytometry Analysis: 2 µg of this antibody detected BLAST-2 (CD23) in one million Raji cells.
Physical properties
Physical form
Storage and Stability
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service