Skip to Content
Merck
All Photos(1)

Key Documents

MABF231

Sigma-Aldrich

Anti-IL1RL1, clone RMST2-2, Azide Free Antibody

clone RMST2-2, from rat

Synonym(s):

Interleukin-1 receptor-like 1, Lymphocyte antigen 84, Protein ST2, Protein T1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€278.00

€278.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€278.00


Estimated to ship on29 April 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

clone

RMST2-2, monoclonal

species reactivity

mouse

technique(s)

activity assay: suitable
flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

IL1RL1, or Interleukin-1 receptor-like 1 protein, which is also known as Lymphocyte antigen 84, Protein ST2, or Protein T1, and encoded by the gene IL1rL1/Ly84/St2/Ste2 is the receptor for IL-33 and its activation recruits MYD88, IRAK1, IRAK4 and TRAF6 transcription factors and promotes the activation of ERK1/MAPK14 and MAPK8. IL1RL1 also is involved in T-helper Cell activation and function. IL1RL1 is localized to the plasma membrane. IL1RL1 is expressed in hematopoietic tissues, macrophages, epithelial and fibroblast cells lines as well as important tissues such as heart and lung. IL1RL1 is a member of the Toll-like receptor superfamily and interestingly plays an important role not in inflammation but in cardiac protection. When IL-33 binds it elicits a cardioprotective signaling response in the tissue and thus helps protect the heart tissue during times of ischemia for instance. Defects in IL1RL1 have been linked to atopic dermatitis and asthma.

Immunogen

Mouse ST-2/IL1RL1.

Application

Activity Assay Analysis: A representative lot from an independent laboratory induces IL-6 production in BMCC cells (Prof. Hisaya Akiba, Jutendo University).
Anti-IL1RL1 antibody, clone RMST2-2, Azide Free is an antibody against IL1RL1 for use in flow cytometry & activity assay.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evalauted by Flow Cytometry in ST2/L5178Y and L5178Y parent cells.

Flow Cytometry Analysis: 1 µg of this antibody detected IL1RL1 in 1X10E6 ST2/L5178Y cells.

Target description

64 kDa calculated

Physical form

Format: Purified
PBS without preservatives.
Protein G Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service