Skip to Content
Merck
All Photos(1)

Key Documents

MABF157D

Sigma-Aldrich

Anti-CD4 (Mouse), redFluor® 710 Antibody, clone RM4-5

clone RM4-5, from rat, redFluor® 710

Synonym(s):

T-cell surface glycoprotein CD4, T-cell differentiation antigen L3T4, T-cell surface antigen T4/Leu-3, CD4, CD4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€128.00
100 μG
€235.00

€128.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€128.00
100 μG
€235.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€128.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

rat

Quality Level

conjugate

redFluor® 710

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM4-5, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Cd4(12504)

General description

The RM4-5 antibody reacts with mouse CD4, a 55 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta2-domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The RM4-5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the RM4-5 antibody and an alternative antibody, Anti-Mouse CD4 clone GK1.5, will “compete” for binding, i.e. RM4-5 antibody is able to block GK1.5 antibody binding to cells. In contrast, RM4-5 antibody does not block the binding of Anti-Mouse CD4 clone RM4-4 to cells.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This rat monoclonal Anti-CD4 Antibody (Mouse), redFluor® 710, clone RM4-5, Cat. No. MABF1577 is validated for use in Flow Cytometry, for the detection of CD4.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected CD4 in one million C57Bl/6 splenocytes.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

redFluor is a registered trademark of Tonbo Biotechnologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service