Skip to Content
Merck
All Photos(2)

Key Documents

MABE927

Sigma-Aldrich

Anti-SAE2 Antibody, clone 7H8.1

clone 7H8.1, from mouse

Synonym(s):

SUMO-activating enzyme subunit 2, Anthracycline-associated resistance ARX, Ubiquitin-like 1-activating enzyme E1B, Ubiquitin-like modifier-activating enzyme 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7H8.1, monoclonal

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBA2(10054)

General description

SUMO-activating enzyme subunit 2, also known as Anthracycline-associated resistance ARX, or Ubiquitin-like 1-activating enzyme E1B, and encoded by the gene name UBA2 or SAE2 or UBLE1B, belongs to the ubiquitin-activating E1 family. SUMOs are a family of small, related proteins (SUMO-1/2/3/4) that can be enzymatically attached to a target protein by a post-translational modification process termed sumoylation, a major regulator of protein function in cellular processes such as nuclear transport, transcriptional regulation, apoptosis and protein stability. This sumoylation is effected by the heterodimeric enzyme SAE1/SAE2 and the SUMO-1-conjugating enzyme Ubch9. The sumoylation pathway mediated by SAE1/SAE2 is distinct from other ubiquitin-like protein (Ubl) pathways.

Immunogen

GST-tagged recombinant protein corresponding to human SAE2.

Application

Anti-SAE2 Antibody, clone 7H8.1 | MABE927 is an antibody against SAE2 Antibody for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected SAE2 in human kidney and rat testis tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in Neuro-2a cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SAE2 in 10 µg of Neuro-2a cell lysate.

Target description

~80 kDa observed. The calculated molecular weight is 72 kDa, however SAE2 has been shown as a ~70-90 kDa band in western blots (Truong, K., et al. (2012). J. Biol. Chem. 287(51):42611-42619).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service