Skip to Content
Merck
All Photos(1)

Key Documents

MABE1805

Sigma-Aldrich

Anti-Rabex-5 Antibody, clone G32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

G32, monoclonal

mol wt

calculated mol wt 56.89 kDa
observed mol wt ~57 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG2aκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RABGEF1(2742)

General description

Rab5 GDP/GTP exchange factor (UniProt: Q9UJ41; also known as RAP1, Rabaptin-5-associated exchange factor for Rab5, Rabex-5) is encoded by the RABGEF1 (also known as RABEX5) gene (Gene ID: 27342) in human. Rabex-5 is a guanine nucleotide exchange factor that catalyzes the exchange of GTP for GDP on the Rab5 subfamily of low molecular weight GTPases. It forms a tight complex with Rabaptin-5 that augments its exchange activity and generates GTP-bound, membrane-associated Rab5, which in turn binds Rabaptin-5 and stabilizes the Rabex-5- Rabaptin-5 complex on endosomes. It is involved in endocytic membrane fusion and membrane trafficking of recycling endosomes. It can also act as a ubiquitin ligase. It binds to ubiquitin (Ub) through two independent ubiquitin-binding domains (UBDs). These UBDs determine a number of properties of Rabex-5, including its coupled monoubiquitination and interaction in vivo with ubiquitinated epidermal growth factor receptors. The first Ub binding region is localized in amino acids 2-49 and is named as RUZ domain (Rabex-5 ubiquitin binding zinc finger). The second Ub binding region is highly conserved and is localized in amino acids 48-74. In this domain Ala58 is shown to be critical for Ub binding and A58G mutagenesis causes significant reduction in its binding to Ub. (Ref.; Penengo, L., et al. (2006). Cell. 124. 1183-1195; Mattera, R., et al. (2006). J. Biol. Chem. 281(10); 6874-6883).

Specificity

Clone G32 is a mouse monoclonal antibody that detects Rabex-5. It targets an epitope within 49 amino acids from the N-terminal region.

Immunogen

GST-tagged full length recombinant human Rabex-5.

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from HeLa cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Rabex-5 in lysates from HeLa cells transfected with either Rabex-5 siRNA or Lamin siRNA. Higher expression was detected in Lamin siRNA transfected HeLa cells.

Tested Applications

Western Blotting Analysis: A representative lot detected Rabex-5 in Western Blotting applications (Penengo, L., et al. (2006). Cell. 124(6): 1183-95).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Rabex-5, clone G32, Cat. No. MABE1805, is a mouse monoclonal antibody that detects Rebex-5 and is tested for use in Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service