Skip to Content
Merck
All Photos(1)

Key Documents

MABE1025

Sigma-Aldrich

Anti-Tankyrase-2 Antibody, clone 2F5.1

clone 2F5.1, from mouse

Synonym(s):

TANK2, Tankyrase-2, ADP-ribosyltransferase diphtheria toxin-like 6, ARTD6, Poly [ADP-ribose] polymerase 5B, TNKS-2, TRF1-interacting ankyrin-related ADP-ribose polymerase 2, Tankyrase II, Tankyrase-like protein, Tankyrase-related protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F5.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TNKS2(80351)

General description

Tankyrase-2 (EC 2.4.2.30; UniProt Q9H2K2; also known as ADP-ribosyltransferase diphtheria toxin-like 6, ARTD6, PARP-5B, Poly [ADP-ribose] polymerase 5B, TANK2, TNKS-2, Tankyrase II, Tankyrase-like protein, Tankyrase-related protein, TRF1-interacting ankyrin-related ADP-ribose polymerase 2) is encoded by the TNKS2 (also known as PARP5B, TANK2, TNKL) gene (Gene ID 80351) in human. Tankyrase-2 belongs to the poly(ADP-ribosylation) polymerase (PARP) family of enzymes that mediate post-translational modification of proteins by poly(ADP-ribosyl)ation, where repeating units of ADP-ribose linked by glycosidic bonds were added to aspartate, glutamate, asparagine, arginine or lysine residues of substrate proteins. Poly(ADP-ribose) (PAR) modification is reversible through the action of poly(ADP-ribose) glycohydrolase (PARG) and ADP-ribosyl protein lyase. Tankyrase-2 and the related Tankyrase-1 share a common domain organization with 15 N-terminal ankyrin-repeats (a.a. 57-776 in TNKS2) involved in substrate recognition, a sterile alpha motif (SAM) domain (a.a. 873-936 in TNKS2) required for dimerization, followed by the C-terminal PARP domain (a.a. 959-1164 in TNKS2). Mice with either Tnks1- or Tnks2-knockout are viable, while double Tnks1-/Tnks2-knockout is lethal at embryonic day 9.5, suggesting a functional redundancy between these two enzymes. Both tankyrases target and suppress Axin2, a well known β-catenin negative regulator. Tankyrase-2 ribosylates Src-binding adapter protein 3BP2, leading to its ubiquitination and proteasomal degradation in macrophages and osteoclasts. Loss of tankyrase-mediated 3BP2 negative regulation underlies the pathogenic mechanism of cherubism, an autosomal dominant disorder affecting cranial bone development.

Specificity

Clone 2F5.1 recognizes an epitope in the region between ANK repeats 6 & 8 of human Tanyrase-2.

Immunogen

Epitope: Region between ANK repeats 6 & 8.
GST-tagged recombinant human Tankyrase-2 fragment from the region between ANK repeats 6 & 8.

Application

Anti-Tankyrase-2 Antibody, clone 2F5.1 is an antibody against Tankyrase-2 for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
General Post-translation Modification

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Tankyrase-2 in 10 µg of Jurkat cell lysate.

Target description

~126 kDa observed. 126.9 kDa (calculated). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service