Skip to Content
Merck
All Photos(1)

Key Documents

MABD129

Sigma-Aldrich

Anti-PRDM1 Antibody, clone 5E7

ascites fluid, clone 5E7, from mouse

Synonym(s):

PR domain zinc finger protein 1, BLIMP-1, Beta-interferon gene positive regulatory domain I-binding factor, PR domain-containing protein 1, Positive regulatory domain I-binding factor 1, PRDI-BF1, PRDI-binding factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€306.00

€306.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5E7, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRDM1(639)

General description

PRDM1/Blimp1 is a repressor of beta-interferon gene expression. In the nucleus, PRDM1 is known to bind specifically with the Beta-interferon gene positive regulatory domain I (PRDI), and to interact with PRMT5. During B-cell differentiation, PRDM1 actively represses the transcription of the gene, c-myc, through binding its promoter and has been deemed a “master regulator” for terminal B-cell differentiation, as it is able to initiate the BCL1 cell developmental cascade. The transcription of this gene is increased upon virus infection.

Immunogen

Purified recombinant fragment of human PRDM1 expressed in E. Coli.

Application

Detect PRDM1 using this mouse monoclonal antibody, Anti-PRDM1 Antibody & clone 5E7 validated for use in western blotting.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in Raji, L1210, and TPH-1 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected PRDM1 in Raji, L1210, and TPH-1 cell lysates.

Optimal working dilutions must be determined by end user.

Target description

~88 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Raji, L1210, and TPH-1 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service