Skip to Content
Merck
All Photos(1)

Documents

MABC1187

Sigma-Aldrich

Anti-Apaf-1 Antibody, clone 18H2

clone 18H2, from rat

Synonym(s):

Apoptotic protease-activating factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

18H2, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... APAF1(317)
mouse ... Apaf1(11783)

General description

Apoptotic protease-activating factor 1 (UniProt: O88879; also known as Apaf-1) is encoded by the Apaf-1 gene (Gene ID: 11783) in murine species. Apaf-1 contains a caspase recruitment domain (CARD), an ATPase domain (NB-ARC), few short helical domains and then several copies of the WD40 repeat domain. It is highly expressed in lung and spleen, weakly in brain and kidney and is not detectable in liver. Under basal conditions it is a monomeric protein, but oligomerizes under apoptotic conditions. The monomeric form is auto-inhibited in a closed conformation through a bound ADP at the nucleotide binding site. Under apoptosis signals ADP is exchanged for ATP and its binding to cytochrome c triggers a conformation change where WD repeat region swings out, which allows the NB-ARC domain to rotate and expose the contact areas for oligomerization. Oligomeric form is a heptameric ring that binds to cytochrome c and ATP and is known as the apoptosome. Oligomeric Apaf-1 mediates the cytochrome c-dependent autocatalytic activation of pro-caspase-9 to mature caspase-9, which then activates effector caspases 3, 6, and 7.

Specificity

Clone 18H2 detects Apaf-1 in human and mouse cells. It targets an epitope within the first 97 amino acids from the N-terminal region containing the CARD domain.

Immunogen

Epitope: N-terminus
Recombinant fragment corresponding to first 97 amino acids from the N-terminal region of murine Apaf-1.

Application

Anti-Apaf-1 Antibody, clone 18H2, Cat. No. MABC1187, is a rat monoclonal antibody that specifically detects Apaf-1. It has been tested for use in Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected Apaf-1 in Western Blotting applications (Scott, C.L., et. al. (2004). J Cell Biol. 164(1):89-96).

Western Blotting Analysis: A representative lot detected Apaf-1 in Western Blotting applications (Marsden, V.S., et. al. (2006). Blood. 107(5):1872-7).

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Apaf-1 in 10 mg of SH-SY5Y cell lysate.




Target description

~130 kDa observed;141 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N V Srikanth Vallabani et al.
Mutagenesis, 34(3), 265-277 (2019-07-05)
Zinc oxide nanoparticles (ZnO NPs) with their wide range of consumer applications in day-to-day life received great attention to evaluate their effects in humans. This study has been attempted to elucidate the DNA damage response mechanism in a dermal model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service