Skip to Content
Merck
All Photos(1)

Documents

MABC1150

Sigma-Aldrich

Anti-MAGEA3 Antibody, clone 57B

clone 57B, from mouse

Synonym(s):

Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3, MAGE-3 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

57B, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MAGEA3(4102)

General description

Melanoma-associated antigen 3 (UniProt: P43357; also known as Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3, MAGE-3 antigen) is encoded by the MAGEA3 (also known as MAGE3) gene (Gene ID: 4102) in human. MAGE-3 is a member of the melanoma-associated antigen gene family of cancer antigens that are expressed in several neoplastic, but not normal, tissues with the exception of testes. In melanoma cells, it enhances viability and serves as an antigen for cytolytic T lymphocytes. MAGEA3 expression is not reported in kidney tumors, leukemias and lymphomas. MAGEA3 expression is normally limited to early developmental stages. It enhances ubiquitin ligase activity of RING-type zinc finger-containing E3 ubiquitin-protein ligases. It is also reported to enhance ubiquitin ligase activity of TRIM28 and stimulate p53/TP53 ubiquitination by TRIM28. It is proposed to act through recruitment and/or stabilization of the Ubl-conjugating enzyme (E2) at the E3:substrate complex.

Specificity

Clone 57B specifically targets a sequence in the N-terminal region of melanoma associated antigen 3 in human testes.

Immunogen

Epitope: domain of:
His-tagged recombinant human melanoma associated antigen 3 containing the polyhistidine tail.

Application

Anti-MAGEA3 Antibody, clone 57B, Cat. No. MABC1150, detects multiple MAGE-A proteins. Validated for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected MAGEA3 in Immunocytochemistry applications. (Hofbauer, G.F., et. al. (1997) Am J Pathol. 151(6):1549-53; Landry, C., et. al. (2000). Int J Cancer. 86(6):835-41; Kocher, T., et. al. (1995). Cancer Res. 55(11):2236-9; Juretic, A., et. al. (2003). Lancet Oncol. 4(2):104-9).

Western Blotting Analysis: A representative lot detected MAGEA3 in Western Blot applications. (Kocher, T., et. al. (1995). Cancer Res. 55(11):2236-9; Rimoldi, D., et. al. (2000). Int J Cancer. 86(5):749-51; SHultz-Thater, E., et. al. (2011). Int J Cancer. 129(5):1137-48).

Immunohistochemistry Analysis: A representative lot detected MAGEA3 in Immunohistochemistry applications. (Hofbauer, G.F., et. al. (1997) Am J Pathol. 151(6):1549-53; Aubry, F., et. al. (2001). Cancer. 92(11):2778-85; Cheville, J.C., et. al. (1999). Mod Pathol. 12(10):974-8; Chitale D.A., et. al. (2005). Mod Pathol. 18(1):119-26; Groeper, C., et. al. (2007). Int J Cancer. 120(2):337-43; Juretic, A., et. al. (2003). Lancet Oncol. 4(2):104-9.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human testis tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected MAGEA3 in human testis tissue.

Target description

34.75 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service