Skip to Content
Merck
All Photos(1)

Documents

MAB748

Sigma-Aldrich

Anti-Salmonella Brazil Antibody, clone 33/204 | MAB748

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

antibody form

purified antibody

Quality Level

clone

33/204
monoclonal

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG2a

Specificity

Salmonella brazil.

Application

ELISA at 1:100 to 1:1,000

IFA

Final working dilutions must be determined by end user.

Physical form

Purified immunoglobulin

Storage and Stability

Maintain at 2-8°C for short term storage. For long term storage, -20°C in convenient aliquots. Avoid repeated freeze/thaw cycles as this reagent may lose activity.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service