Skip to Content
Merck
All Photos(1)

Key Documents

MAB5596

Sigma-Aldrich

Anti-HCN2 Antibody

culture supernatant, Chemicon®

Synonym(s):

Hyperpolarization-activated and cyclic nucleotide-gated channel 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€347.00

€347.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€347.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Hcn2(15166)
rat ... Hcn2(114244)

Specificity

HCN2 (Hyperpolarization-activated and cyclic nucleotide-gated channel 2). The antibody recognizes both the glycosyated and deglycosylated form of HCN2. The antibody recognizes a weak band at ~98 kDa and a stronger band at ~108 kDa in glycosylated protein from a retinal membrane preparation.

Immunogen

Synthetic peptide from rat HCN2.

Application

Anti-HCN2 Antibody is an antibody against HCN2 for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot on rat and mouse retina. The antibody recognizes a weak band at ~98 kDa and a stronger band at ~108 kDa in glycosylated protein from a retinal membrane preparation.

Immunohistochemistry on rat and mouse retina.

Optimal working dilutions must be determined by end user.

Physical form

Cultured supernatant containing 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joon Ha Park et al.
International journal of molecular medicine, 44(5), 1801-1810 (2019-10-02)
Hyperpolarization‑activated cyclic nucleotide‑gated (HCN) channels have been known to participate in the regulation of neuronal excitability, synaptic transmission and long‑term potentiation in the hippocampus. The present study investigated transient ischemia‑induced changes of HCN1 and HCN2 expressions in the Cornu Ammonis
Stuart M Cain et al.
Pflugers Archiv : European journal of physiology, 467(6), 1367-1382 (2014-06-24)
Burst-firing in distinct subsets of thalamic relay (TR) neurons is thought to be a key requirement for the propagation of absence seizures. However, in the well-regarded Genetic Absence Epilepsy Rats from Strasbourg (GAERS) model as yet there has been no

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service