Skip to Content
Merck
All Photos(1)

Key Documents

MAB3314

Sigma-Aldrich

Anti-MMP-16 Antibody, clone 117-10C6

clone 117-10C6, Chemicon®, from mouse

Synonym(s):

MT3-MMP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

117-10C6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MMP16(4325)

Specificity

The antibody specifically reacts with human MT3-MMP. This is a purified mouse monoclonal antibody to an oligopeptide of EEVPYSELENGKRD (residue 168-181) on human membrane-type 3 matrix metalloproteinase (human MT3-MMP).

Immunogen

This antibody was made to an oligopeptide of EEVPYSELENGKRD (residue 168-181) on human membrane-type 3 matrix metalloproteinase (human MT3-MMP).

Application

Anti-MMP-16 Antibody, clone 117-10C6 detects level of MMP-16 & has been published & validated for use in WB.
Immunoblotting

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cameron P Bracken et al.
The EMBO journal, 33(18), 2040-2056 (2014-07-30)
The microRNAs of the miR-200 family maintain the central characteristics of epithelia and inhibit tumor cell motility and invasiveness. Using the Ago-HITS-CLIP technology for transcriptome-wide identification of direct microRNA targets in living cells, along with extensive validation to verify the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service