Skip to Content
Merck
All Photos(2)

Key Documents

LUXRGTDIL

Millipore

mPAGE® Lux Bis-Tris Diluent for mPAGE® Lux Curing Station

greener alternative

Diluent for use with Resolving Solution for MPAGE® Lux Curing Station

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NC.01
Pricing and availability is not currently available.

Product Name

mPAGE® Lux Bis-Tris Diluent for mPAGE® Lux Curing Station,

shelf life

12 mo.

Quality Level

packaging

1 ea of 1 ea

manufacturer/tradename

Millipore

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable
immunoblotting: suitable
western blot: suitable

L × W × H

7.5 cm × 7.5 cm × 13.4 cm

greener alternative category

storage temp.

2-8°C

General description

Use the mPAGE® Lux Bis-Tris Diluent with mPAGE® Lux Curing Station, mPAGE® Lux Resolving Solution and mPAGE® Lux Stacking Solution for making polyacrylamide gels. It offers rapid and simple gel curing process for handcasting mini protein gels. A mini gel is cured in 1.5min and overall casting process can be less than 3min.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service