Skip to Content
Merck
All Photos(1)

Key Documents

HTS086RTA

Sigma-Aldrich

Ready-to-Assay H2 Histamine receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Pricing and availability is not currently available.

Product Name

Ready-to-Assay H2 Histamine receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells, resuspends them in media, and allows them to recover for 24 hours. The following day the cells are counted, plated, and assayed for calcium response.

The histamine H2 receptor is a widely distributed G-protein coupled receptor, which is important in regulating a host of physiologic actions extending from gastric acid secretion to gastrointestinal motility (Del Valle and Gantz, 1997). Histamine mediated activation of its receptor leads to signaling through both adenylate cyclase and phosphoinositide /protein kinase C second messenger systems (Hill, 1990). Antagonists for this receptor have proven to be effective therapy for acid peptic disorders of the GI tract. Certain antagonists are used in the treatment of neuropsychiatric and neurological diseases e.g. schizophrenia, Alzheimer′s disease and Parkinson′s disease. Millipore′s cloned human H2-expressing cell line is made in the Chem-2 host, which supports high levels of recombinant H2 expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at H2.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-2

Application

Human H2 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: H2
  • Target Sub-Family: Histamine

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service