Skip to Content
Merck
All Photos(2)

Key Documents

HTS080M

Sigma-Aldrich

ChemiSCREEN Human NK1 Tachykinin Receptor Membrane Preparation

Human NK1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Full-length human TACR1 cDNA encoding NK1
The tachykinin peptide family in mammals comprises three peptides, substance P, neurokinin A and neurokinin B, which bind to the tachykinin receptor family of GPCRs, NK1, NK2 and NK3 (Severini et al., 2002). Tachykinins have prominent activity in the GI system, in which they stimulate intestinal contraction and salivation. These effects are mediated by NK1 and NK2, and an antagonist of NK1, aprepitant, is used for treatment of chemotherapy-induced emesis (Rupniak and Kramer, 1999). The NK1 tachykinin receptor is expressed in brain, and is thought to be involved in depression and nociception (Saria, 1999). Although NK1 appears to promote nociception in animal models, the significance of this activity is controversial, as antagonists of NK1 have not proven efficacious in pain relief in humans (Hill, 2000). Chemicon′s NK1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of NK1 interactions with Substance P. The membrane preparations exhibit a Kd of 0.28-0.4 nM for [125I]-Substance P. With 10 µg/well NK1 Membrane Prep and 0.25 nM [125I]-Substance P, a greater than 5-fold signal-to-background ratio was obtained.

Application

Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: NK1
Target Sub-Family: Tackykinin /neurokinin

Quality



Signal:background and specific binding values obtained in a competition binding assay with varying amounts of NK1 membrane prep:
10 µg/well
Signal:Background 7.17
Specific Binding (cpm) 2210


SPECIFICATIONS: 1 unit = 10 µg
Bmax : 0.38 pmol/mg
Kd: 0.34 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-Substance P (Perkin Elmer#:NEX190 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 5-fold signal:background with 125I labeled Substance P at 0.25 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 mL packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hill
Trends in pharmacological sciences, 21(7), 244-246 (2000-06-29)
Tachykinin NK1 receptor antagonists have failed to exhibit efficacy in clinical trials of a variety of clinical pain states. By contrast, in preclinical studies in animals NK1 receptor antagonists have been shown to attenuate nociceptive responses sensitized by inflammation or
N M Rupniak et al.
Trends in pharmacological sciences, 20(12), 485-490 (2000-02-12)
The development of small-molecule antagonists of the substance P (SP)-preferring tachykinin NK1 receptor during the past decade represents an important opportunity to exploit these molecules as novel therapeutic agents. On the basis of its anatomical localization and function, SP has
Cinzia Severini et al.
Pharmacological reviews, 54(2), 285-322 (2002-05-31)
The tachykinin peptide family certainly represents one of the largest peptide families described in the animal organism. So far, more than 40 tachykinins have been isolated from invertebrate (insects, worms, and molluscs), protochordate, and vertebrate (skin, gastrointestinal tract, peripheral and
A Saria
European journal of pharmacology, 375(1-3), 51-60 (1999-08-12)
After its discovery in 1931, substance P (SP) remained the only mammalian member of the family of tachykinin peptides for several decades. Tachykinins thus refer to peptides sharing the common C-terminal amino acid sequence Phe-X-Gly-Leu-Met x NH2. In recent years

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service