Skip to Content
Merck
All Photos(1)

Key Documents

GM05/4A11-6410

Millipore

NovAseptic® GMP 50 Mixer Drive Unit

Drive Unit Compatible with Speed Counter and 60 mm Shaft

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151800

Quality Level

parameter

max. pressure at 25 °C (atmospheric pressure)
0-40 °C max. temp. (32-104 °F)

General description

NovAseptic® mixers offer a complete solution for mixing throughout the
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.

Specificity

General Motor Data:Voltage: 230/400V AC - 50 Hz and 460 V - 60 HzNumber of Poles: 2Degree of Protection: IP 55Color: Green, complies with the Code of Federal Regulations (FDA), US 21 CFR 170 - 199.Oil: Synthetic, complies with the Code of Federal Regulations (FDA) 21 CFRCertification: CE / CSA/ ULThermistor 1 (PTC, 155 °C)

Application

NovAseptic® mixers offer a complete solution for mixing throughout the
process line and are designed for a wide variety of mixing applications in the
pharmaceutical and biotechnology industries.

Components

The drive unit is packaged in a closed box.

Legal Information

NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service