GF037
Thrombopoietin
TPO (Thrombopoietin, Mpl-ligand, MGDF) stimulates the proliferation & maturation of megakaryocytes & promotes increased circulating levels of platelets in vivo.
Synonym(s):
TPO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
Assay
>98% (SDS-PAGE and HPLC)
specific activity
≥1x10(e6) U/mg
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 ng/mg Endotoxin (of human TPO; 1 EU/μg)
input
sample type hematopoietic stem cell(s)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... TPO(7173)
General description
Product Source: E.coli
TPO (Thrombopoietin, Mpl-ligand, MGDF) stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo. Chemicon′s Human TPO is a soluble 18.5 kDa protein containing 174 amino acid residues which comprises the receptor binding domain of the Mpl-ligand protein.
Biochem/physiol Actions
The ED50 was determined by the dose-dependent stimulation of the proliferation of M07e cells is ≤1.0 ng/mL
Physical form
Lyophilized protein from 5mM Tris, pH 7.6, with no additives.
Storage and Stability
The lyophilized powder, though stable at room temperature, is best stored at -20°C. Reconstitute in 5 mM Tris, pH 8.0 to a concentration of 0.5-1.0 mg/mL. This solution can be diluted into other aqueous buffers and stored at 4ºC for 1 week or -20ºC for future use. Reconstituted TPO should be stored in working aliquots at -20°C.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service