Skip to Content
Merck
All Photos(1)

Key Documents

EZHNPY

Millipore

Human Neuropeptide Y ELISA Kit

measures and quantifies NPY levels in 50 μL serum or plasma

Synonym(s):

Pro-neuropeptide Y

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
€1,030.00

€1,030.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
96 WELLS
€1,030.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

€1,030.00


Estimated to ship on23 May 2025


Request a Bulk Order

Product Name

Human Neuropeptide Y (NPY) ELISA, This Human Neuropeptide Y (NPY) ELISA is used to measure & quantify NPY levels in Metabolism & Neuroscience research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (4hr assay)

assay range

accuracy: 94-99%
sensitivity: 3 pg/mL
(50 μl sample size)

standard curve range: 5-1000 pg/mL
inter-assay cv: 3-14%
intra-assay cv: 2-5%

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Millipore is pleased to offer our new 96-well ELISA NPY kit. This kit requires no sample extraction and supports our commitment to the study of various metabolic disease markers.
Neuropeptide Y (NPY) is a 36-amino acid peptide found primarily in the brain. It has been associated with a number of physiologic processes, including the regulation of energy, balance, memory and learning. NPY is secreted by the hypothalamus, and in addition to increasing appetite, it increases the proportion of energy stored as fat and blocks signals to the brain. NPY is closely related to other peptide hormones such as pancreatic polypeptide, PYY and correlates nicely to Leptin levels.

Specificity

Cross Reactivty
Refer to kit protocol for details

Application

Research Category
Neuroscience
This assay requires 50 µl of sample and is performed at room temperature as a 4 hour assay.
Used to detect/quantify: NPY

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I would like to have a protocol (pdf) for EZHNPY-25K. Where can I download it?

    1 answer
    1. Full product information, including the protocol, can be found at the link below. See the Documentation section under Protocols:
      https://www.emdmillipore.com/product/Human-Neuropeptide-Y-NPY-ELISA,MM_NF-EZHNPY-25K#documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service