Skip to Content
Merck
All Photos(1)

Key Documents

EZFLA4FT1

Millipore

EZ-Fluo Functional Test Report

Verification Report for Core Functions in A4 Format

Synonym(s):

Rapid Microbial Detection System Verification Report

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€337.00

€337.00


Estimated to ship on25 May 2025


Request more information

Select a Size

Change View
1 UNIT
€337.00

About This Item

UNSPSC Code:
41116500
NACRES:
ND.05

€337.00


Estimated to ship on25 May 2025


Request more information

manufacturer/tradename

EZ-Fluo

compatibility

for use with EZ-Fluo Rapid Detection System.

General description

EZ-Fluo Functional Test Report is designed as a light guide for users to perform a verification exercise of the core functions of the equipment after purchase and before starting with routine testing of samples. EZ-Fluo system is a rapid microbial detection system which is designed to detect live microorganism present in test samples. The EZ-Fluo system can detect and quantify microbial contamination in any filterable sample.
The verification protocol in A4 format ensures optimal performance of the EZ-Fluo system.

Application

EZ-Fluo Functional Test Report in A4 format is used to perform verification of core functions of the equipment.

Legal Information

EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service