DR1002
PhosphoDetect Anti-ATM (pSer¹⁹⁸¹) Mouse mAb (10H11.E12)
liquid, clone 10H11.E12, Calbiochem®
Synonym(s):
Anti-Ataxia Telangiectasia, Phospho-Specific (Ser¹⁹⁸¹)
About This Item
biological source
mouse
Quality Level
antibody product type
primary antibodies
clone
10H11.E12, monoclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
human, mouse
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
General description
Immunogen
Application
Immunoblotting (0.5 g/ml)
Immunocytochemistry (1.5 g/ml)
Packaging
Warning
Physical form
Reconstitution
Analysis Note
HeLa cells treated with camptothecin (Cat. No. 208925)
Other Notes
Abraham, R.T. 2003. Genes Dev.15, 2177.
Bakkenist, C.J. and Kastan, M.B. 2003. Nature421, 499.
Shiloh, Y. 2003. Nat. Rev. Cancer3, 155.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service