Skip to Content
Merck
All Photos(1)

Key Documents

CX2065

Sigma-Aldrich

Cresyl Violet Acetate

HARLECO® For the staining of nuclei and Nissl bodies

Synonym(s):

Cresyl Violet Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
MDL number:
UNSPSC Code:
41116130
EC Index Number:
234-043-3

Quality Level

form

powder or crystals

pH

3 (20 °C, 10 g/L in H2O)

mp

140-143 °C

solubility

70 g/L

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C16H12N3O.C2H4O2/c17-9-5-6-13-14(7-9)20-15-8-12(18)10-3-1-2-4-11(10)16(15)19-13;1-2(3)4/h1-8H,17-18H2;1H3,(H,3,4)/q+1;/p-1

InChI key

XKOCOMKJPWEOHX-UHFFFAOYSA-M

Application

For In-Vitro Diagnostic Use.
For Professional Use.
HARLECO® For the staining of nuclei and Nissl bodies

Analysis Note

Absorbance at maximum0.50AU minAbsorbance maximum wavelength596-599nmAbsorbance ratio; (P-15/P+15)0.94-1.20Assay (TiCl3 titration)65% minColorGreen to blackFormPowder and/or crystals

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service