Skip to Content
Merck
All Photos(1)

Documents

CX1055

Supelco

Chloroform

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Trichloromethane, Chloroform, Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-663-8

Agency

meets USP/NF

Quality Level

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8% chloroform + ethanol basis

form

liquid

impurities

≤0.03% Water (H2O)
≤0.05 ppm Lead (Pb)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8.7 g/L

density

1.492 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CX1055P-4 is packaged in poly coated glass bottles
Stabilized with Ethanol.
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Acetone and Aldehydes (about 0.005%)To pass testAcid and chlorideTo pass testAssay (CHCl3 + C2H5OH)99.8% minColor (APHA)10 maxDensity at 25°C1.465-1.476g/mLEvaporation residue0.001% maxFormClear liquidFree chlorine (Cl)To pass testLead (Pb)0.05ppm maxSubstances darkened by sulfuric acidTo pass testSuitability for determination with dithizoneTo pass testWater (H2O)0.03% max

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service