Skip to Content
Merck
All Photos(1)

Documents

CX0395

Supelco

Carbon Disulfide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Carbon Disulfide, Carbon Bisulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12191602
EC Index Number:
200-843-6

Agency

meets USP/NF

Quality Level

vapor pressure

398 hPa ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

impurities

≤0.05% Water (H2O)

evapn. residue

≤0.002%

color

APHA: ≤10

bp

46.5 °C/1013 hPa

mp

-111.6 °C

transition temp

flash point -30 °C

solubility

2.1 g/L

density

1.26 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CX0395-6 and CX0395-5 are packaged in poly coated glass bottles
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (GC): 99.9% min
Color (APHA): 10 max
Evaporation residue: 0.002% max
Form: Clear liquid
Hydrogen sulfide (H2S): To pass test (about 1.5 ppm)
Sulfur dioxide (SO2): To pass test (about 2.5 ppm)
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service